Learn More
Labconco™ 5 ft. Purifier Logic+ Class II Type A2 Biosafety Cabinet with 12 in. Sash Opening
5 ft. Purifier Logic+ Class II Type A2 Biosafety Cabinet with 12 in. Sash Opening creates a particulate-free environment for small animal research activities such as cage changing, weighing, anesthesia delivery and surgical procedures.
$15260.00 - $16800.00
Specifications
Certifications/Compliance NSF/ANSI Standard 49, UL Standard 61010-1, CAN/CSA 22.2 No. 61010-1, CE Conformity Marking (230 V models), ISO 5 conditions, ISO 14644-1 and 2, ADA-compliant operation
Item Description Class II Type A2 Biosafety Cabinet
View More Specs
Products 20
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
30-252-0021
View Documents
Labconco™
302520021
Japan 100 V, 50/60 Hz, 12 A Biosafety Cabinet only
Each for $15,260.00
Add to cart
30-252-0031
View Documents
Labconco™
302520031
Schuko 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $15,260.00
Add to cart
30-252-0041
View Documents
Labconco™
302520041
China/Australia 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $15,260.00
Add to cart
30-252-0051
View Documents
Labconco™
302520051
British (UK) 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $15,260.00
Add to cart
30-252-0071
View Documents
Labconco™
302520071
India 230 V, 50/60 Hz, 6 A Biosafety Cabinet only
Each for $15,260.00
Add to cart
30-252-0121
View Documents
Labconco™
302520121
Japan 100 V, 50/60 Hz, 12 A Base Stand
Each for $16,060.00
Add to cart
30-252-0131
View Documents
Labconco™
302520131
Schuko 230 V, 50/60 Hz, 6 A Base Stand
Each for $16,060.00
Add to cart
30-252-0141
View Documents
Labconco™
302520141
China/Australia 230 V, 50/60 Hz, 6 A Base Stand
Each for $16,060.00
Add to cart
30-252-0151
View Documents
Labconco™
302520151
British (UK) 230 V, 50/60 Hz, 6 A Base Stand
Each for $16,060.00
Add to cart
30-252-0171
View Documents
Labconco™
302520171
India 230 V, 50/60 Hz, 6 A Base Stand
Each for $16,060.00
Add to cart
30-252-1021
View Documents
Labconco™
302521021
Japan 100 V, 50/60 Hz, 12 A Accessory Pk.
Each for $16,000.00
Add to cart
30-252-1031
View Documents
Labconco™
302521031
Schuko 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $16,000.00
Add to cart
30-252-1041
View Documents
Labconco™
302521041
China/Australia 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $16,000.00
Add to cart
30-252-1051
View Documents
Labconco™
302521051
British (UK) 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $16,000.00
Add to cart
30-252-1071
View Documents
Labconco™
302521071
India 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $16,000.00
Add to cart
30-252-1121
View Documents
Labconco™
302521121
Japan 100 V, 50/60 Hz, 12 A Base Stand and Accessory Pk.
Each for $16,800.00
Add to cart
30-252-1131
View Documents
Labconco™
302521131
Schuko 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $16,800.00
Add to cart
30-252-1141
View Documents
Labconco™
302521141
China/Australia 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $16,800.00
Add to cart
30-252-1151
View Documents
Labconco™
302521151
British (UK) 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $16,800.00
Add to cart
30-252-1171
View Documents
Labconco™
302521171
India 230 V, 50/60 Hz, 6 A Base Stand and Accessory Pk.
Each for $16,800.00
Add to cart
Description
Description
  • Purifier Logic+ Class II A2 Biosafety Cabinets provide personnel, product and environmental protection and are suitable for work in all biosafety levels
  • Designed to discharge HEPA-filtered exhaust air directly into the laboratory, or into an exhaust system through the optional Canopy Connection
  • Some models have base stands and/or an Accessory Package (UV light, service fixture(s) and Vacu-Pass™ Cord and Cable Portal) included
  • Inclination™ Technology Suite that includes MyLogic™ operating system with eye-level color display, 10° angled sash, patented curved air grille with built in armrest, plus fully accessible controls, outlets, and fixtures
  • MyLogic is easy to use, displaying the BSC's status with clear messaging and graphics that notify users of filter life, alerts, and alarms
  • Energy-efficient ECM blower powered by Labconco’s exclusive Constant Airflow Profile™ (CAP) technology
Specifications
Specifications
NSF/ANSI Standard 49, UL Standard 61010-1, CAN/CSA 22.2 No. 61010-1, CE Conformity Marking (230 V models), ISO 5 conditions, ISO 14644-1 and 2, ADA-compliant operation
Class II Type A2 Biosafety Cabinet