Alfa Aesar™ 5-Hexadecanone, 99%

Manufacturer: Alfa Aesar™ B2488114

Quantity 25g

 View more versions of this product

Catalog No. AAB2488114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 41903-81-5
    Molecular Formula C16H32O
    Formula Weight 240.43
    MDL Number MFCD00051550
    Synonym n-Butyl n-undecyl ketone
    Chemical Name or Material 5-Hexadecanone

    Specifications

    Formula Weight 240.43
    MDL Number MFCD00051550
    CAS 41903-81-5
    Quantity 25g
    Molecular Formula C16H32O
    Synonym n-Butyl n-undecyl ketone
    Melting Point 36° to 38°C
    Chemical Name or Material 5-Hexadecanone
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit