Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Hexadecanone, 99%

Click to view available options
Quantity:
5g
25g
100g
41903-81-5
C16H32O
240.431
MFCD00051550
GASJOQPNNSMIFF-UHFFFAOYSA-N
5-hexadecanone,n-butyl n-undecyl ketone,acmc-20am37
98680
hexadecan-5-one
CCCCCCCCCCCC(=O)CCCC
41903-81-5
C16H32O
240.431
MFCD00051550
GASJOQPNNSMIFF-UHFFFAOYSA-N
5-hexadecanone,n-butyl n-undecyl ketone,acmc-20am37
98680
hexadecan-5-one
CCCCCCCCCCCC(=O)CCCC
Quantity 100g
Melting Point 36°C to 38°C
Formula Weight 240.43
Percent Purity 99%
Chemical Name or Material 5-Hexadecanone