Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexen-1-ol, 99%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
100mL
Packaging:
Glass bottle
821-41-0
C6H12O
100.16
UIZVMOZAXAMASY-UHFFFAOYSA-N
5-hexen-1-ol,1-hexen-6-ol,5-hexenol,unii-57pd1rf6g7,5-hexene-1-ol,5-hexanol,hex-1-en-6-ol,acmc-209pn9,ho ch2 4ch=ch2
69963
hex-5-en-1-ol
C=CCCCCO
821-41-0
C6H12O
100.16
UIZVMOZAXAMASY-UHFFFAOYSA-N
5-hexen-1-ol,1-hexen-6-ol,5-hexenol,unii-57pd1rf6g7,5-hexene-1-ol,5-hexanol,hex-1-en-6-ol,acmc-209pn9,ho ch2 4ch=ch2
69963
hex-5-en-1-ol
C=CCCCCO
Infrared Spectrum Authentic
Refractive Index 1.4340 to 1.4360
Quantity 5mL
Specific Gravity 0.8725
Density 0.8725g/mL
Color Colorless
Boiling Point 152.0°C to 155.0°C
Flash Point 47°C
Packaging Glass bottle
Assay Percent Range 98.5% min. (GC)
Linear Formula H2C=CH(CH2)4OH
Beilstein 01,IV,2137
Solubility Information Solubility in water: 18.6g/L (20°C).
Formula Weight 100.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 5-Hexen-1-ol
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 212-477-4