Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexynoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
53293-00-8
C6H8O2
112.13
MFCD00066346
VPFMEXRVUOPYRG-UHFFFAOYSA-N
5-hexynoic acid,5-hexyn-1-oic acid,4-ethynylbutyric acid,acmc-1an4h
143036
CHEBI:73511
hex-5-ynoic acid
C#CCCCC(=O)O
53293-00-8
C6H8O2
112.13
MFCD00066346
VPFMEXRVUOPYRG-UHFFFAOYSA-N
5-hexynoic acid,5-hexyn-1-oic acid,4-ethynylbutyric acid,acmc-1an4h
143036
CHEBI:73511
hex-5-ynoic acid
C#CCCCC(=O)O
Density 1.0160g/mL
Infrared Spectrum Authentic
Boiling Point 224.0°C to 225.0°C
Flash Point 110°C
Specific Gravity 1.016
Packaging Glass bottle
Refractive Index 1.4480 to 4.4500
Quantity 1g
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH≡C(CH2)3COOH
Formula Weight 112.13
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 5-Hexynoic acid
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes serious eye damage.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger