Learn More
5-hydroxymethylcytosine Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA540097
Catalog No. PA540097
Add to cart
Description
Description
5-hydroxymethylcytosine (5-hmC) has been recently discovered in mammalian DNA. 5-hmC results from the enzymatic conversion of 5-methylcytosine into 5-hydroxymethylcytosine by the TET family of oxygenases. Initially, the 5-hmC base had been identified in Purkinje neurons, in granule cells and embryonic stem cells where it is present at high levels (up to 0,6% of total nucleotides in Purkinje cells). A recent report indicates that 5-hmC is also abundant in brain tissue, especially in areas that are associated with higher cognitive functions.Early evidence suggests that 5-hmC may represent an intermediate in a new pathway which demethylates DNA, converting 5-mC to cytosine. Although its precise role has still to be shown, recent results indicate, however, that 5-hmC may play important roles distinct from 5-mC. This may open up entirely new perspectives in epigenetic studies.
Specifications
Specifications
5-hydroxymethylcytosine
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
50 μg
-20°C or -80°C if preferred
Primary
Human, Mouse
ELISA, Immunoprecipitation, Peptide Array
3.5 mg/mL
PBS with 0.05% Proclin 300, 0.05% sodium azide; pH 7.4
5hmc; 5-hmc; 5-HydroxymethylCytidine; C5H7N3O2
Human KLH-conjugated 5-hydroxymethylcytosine.
Protein G
RUO
Antibody
Polyclonal