Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT3B Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $320.71

Specifications

Antigen 5-HT3B
Dilution Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125618
View Documents
Novus Biologicals
NBP31035825UL
25 μg
Each for $106.19
Add to cart
 
NB125617
View Documents
Novus Biologicals
NBP310358100UL
100 μg
Each for $320.71
Add to cart
 
Description

Description

5-HT3B Polyclonal antibody specifically detects 5-HT3B in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

5-HT3B
Western Blot, Immunohistochemistry, Immunofluorescence
Purified
RUO
PBS buffer, 2% sucrose
9177
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Unconjugated
Rabbit
Human, Mouse
5-HT3-B, 5-HT3B5-hydroxytryptamine receptor 3B, 5-hydroxytryptamine (serotonin) receptor 3B, 5-hydroxytryptamine 3 receptor B subunit, Serotonin receptor 3B, serotonin-gated ion channel subunit
The immunogen is a synthetic peptide directed towards the N terminal region of human 5-HT3B (NP_006019). Peptide sequence PLWACILVAAGILATDTHHPQDSALYHLSKQLLQKYHKEVRPVYNWTKAT
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit