Learn More
5-Hydroxy-1,4-naphthoquinone, 99%, Thermo Scientific Chemicals
Description
5-Hydroxy-1,4-naphthoquinone is used as a natural dye in cloth, fabrics, wool and ink. It is a coloring agent for food and cosmetics. It is involved in the synthesis of poly(hydroxyl-1,4-naphthoquinone) stabilized gold nanoparticles (AuNQ NPs), which is used for nonenzymatic electrochemical detection of glucose.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
5-Hydroxy-1,4-naphthoquinone is used as a natural dye in cloth, fabrics, wool and ink. It is a coloring agent for food and cosmetics. It is involved in the synthesis of poly(hydroxyl-1,4-naphthoquinone) stabilized gold nanoparticles (AuNQ NPs), which is used for nonenzymatic electrochemical detection of glucose.
Solubility
Soluble in alcohol, chloroform, dimethylsulfoxide, ether and aqueous solution of alkalies. Slightly soluble in water.
Notes
Light sensitive. Incompatible with strong bases, strong reducing agents and strong oxidizing agents.
Specifications
Melting Point | 161°C to 163°C |
Quantity | 5 g |
UN Number | UN2811 |
Sensitivity | Light sensitive |
Merck Index | 14,5629 |
Solubility Information | Soluble in alcohol,chloroform,dimethylsulfoxide,ether and aqueous solution of alkalies. Slightly soluble in water. |
Formula Weight | 174.16 |
Percent Purity | 99% |
Chemical Name or Material | 5-Hydroxy-1,4-naphthoquinone |
Safety and Handling
Hazard Category | H301-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 207-567-5 |
RTECSNumber | QJ5775000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.