Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-1-tetralone, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
28315-93-7
C10H10O2
162.188
MFCD00001693
YPPZCRZRQHFRBH-UHFFFAOYSA-N
5-hydroxy-1-tetralone,1-tetralon-5-ol,5-hydroxy-3,4-dihydronaphthalen-1 2h-one,5-hydroy-1-tetraione,1 2h-naphthalenone, 3,4-dihydro-5-hydroxy,5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one,5-hydroxytetralone,3,4-dihydro-5-hydroxy-1 2h-naphthalenone,1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one,5-hydroxy-2,3,4-trihydronaphthalen-1-one
119921
5-hydroxy-3,4-dihydro-2H-naphthalen-1-one
C1CC2=C(C=CC=C2O)C(=O)C1
28315-93-7
C10H10O2
162.188
MFCD00001693
YPPZCRZRQHFRBH-UHFFFAOYSA-N
5-hydroxy-1-tetralone,1-tetralon-5-ol,5-hydroxy-3,4-dihydronaphthalen-1 2h-one,5-hydroy-1-tetraione,1 2h-naphthalenone, 3,4-dihydro-5-hydroxy,5-hydroxy-1,2,3,4-tetrahydronaphthalen-1-one,5-hydroxytetralone,3,4-dihydro-5-hydroxy-1 2h-naphthalenone,1,2,3,4-tetrahydro-5-hydroxynaphthalen-1-one,5-hydroxy-2,3,4-trihydronaphthalen-1-one
119921
5-hydroxy-3,4-dihydro-2H-naphthalen-1-one
C1CC2=C(C=CC=C2O)C(=O)C1
Color Beige to Yellow or Pink
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 207°C to 210°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 08, III, 820
Formula Weight 162.19
Physical Form Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Hydroxy-1-tetralone, 99%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 248-958-0