Alfa Aesar™ 5-Hydroxy-2-adamantone, 98%

Manufacturer: Alfa Aesar™ H5595006

Quantity 5g

 View more versions of this product

Catalog No. AAH5595006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20098-14-0
    Molecular Formula C10H14O2
    Formula Weight 166.22
    MDL Number MFCD00192211
    Chemical Name or Material 5-Hydroxy-2-adamantone

    Specifications

    Formula Weight 166.22
    CAS 20098-14-0
    Melting Point >300°C
    Quantity 5g
    Molecular Formula C10H14O2
    MDL Number MFCD00192211
    Chemical Name or Material 5-Hydroxy-2-adamantone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit