Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-2-coumaranone, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
2688-48-4
C8H6O3
150.133
MFCD00005857
POUITAHNNRJWMA-UHFFFAOYSA-N
5-hydroxybenzofuran-2 3h-one,5-hydroxy-2-coumaranone,homogentisic acid gamma-lactone,5-hydroxybenzofuran-2-one,2,3-dihydro-5-hydroxybenzofuran-2-one,5-hydroxy-2 3h-benzofuranone,2 3h-benzofuranone, 5-hydroxy,homogentisic acid,a-lactone,5-hydroxy-3-hydrobenzo b furan-2-one,homogentisic lactone
75898
5-hydroxy-3H-1-benzofuran-2-one
C1C2=C(C=CC(=C2)O)OC1=O
2688-48-4
C8H6O3
150.133
MFCD00005857
POUITAHNNRJWMA-UHFFFAOYSA-N
5-hydroxybenzofuran-2 3h-one,5-hydroxy-2-coumaranone,homogentisic acid gamma-lactone,5-hydroxybenzofuran-2-one,2,3-dihydro-5-hydroxybenzofuran-2-one,5-hydroxy-2 3h-benzofuranone,2 3h-benzofuranone, 5-hydroxy,homogentisic acid,a-lactone,5-hydroxy-3-hydrobenzo b furan-2-one,homogentisic lactone
75898
5-hydroxy-3H-1-benzofuran-2-one
C1C2=C(C=CC(=C2)O)OC1=O
Quantity 1g
Packaging Glass bottle
Color Beige
Melting Point 190°C to 195°C
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 18, 17
Formula Weight 150.13
Physical Form Fine Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 5-Hydroxy-2-coumaranone
Show More Show Less