Alfa Aesar™ 5-Hydroxy-2-methylindole, 98%

Manufacturer: Alfa Aesar™ H3558009

Quantity 10g

 View more versions of this product

Catalog No. AAH3558009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13314-85-7
    Formula Weight 147.18
    MDL Number MFCD00022714
    Chemical Name or Material 5-Hydroxy-2-methylindole

    Specifications

    Formula Weight 147.18
    Melting Point 130° to 132°C
    Storage Note 1 2° to 8°C
    Sensitivity Light Sensitive
    Color Brown
    Physical Form Solid
    MDL Number MFCD00022714
    CAS 13314-85-7
    Quantity 10g
    Chemical Name or Material 5-Hydroxy-2-methylindole
    Assay 98%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit