Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Hydroxy-2-methylpyridine, 99%

Click to view available options
Quantity:
5g
25g
100g
1121-78-4
C6H7NO
109.128
MFCD00006339
DHLUJPLHLZJUBW-UHFFFAOYSA-N
5-hydroxy-2-methylpyridine,2-methyl-5-hydroxypyridine,3-hydroxy-6-methylpyridine,6-methyl-3-pyridinol,3-pyridinol, 6-methyl,6-methyl-3-hydroxypyridine,5-hydroxy-2-picoline,unii-uke16jp1jr,2-methyl-5-pyridinol,uke16jp1jr
14275
6-methylpyridin-3-ol
CC1=NC=C(C=C1)O
1121-78-4
C6H7NO
109.128
MFCD00006339
DHLUJPLHLZJUBW-UHFFFAOYSA-N
5-hydroxy-2-methylpyridine,2-methyl-5-hydroxypyridine,3-hydroxy-6-methylpyridine,6-methyl-3-pyridinol,3-pyridinol, 6-methyl,6-methyl-3-hydroxypyridine,5-hydroxy-2-picoline,unii-uke16jp1jr,2-methyl-5-pyridinol,uke16jp1jr
14275
6-methylpyridin-3-ol
CC1=NC=C(C=C1)O
Melting Point 167°C to 171°C
Odor Odorless
Quantity 100g
Beilstein 107077
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 5-Hydroxy-2-methylpyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : UU7714900