Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-2-nitrobenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
42454-06-8
C7H5NO4
167.12
XLYPHUGUKGMURE-UHFFFAOYSA-N
3-formyl-4-nitrophenol,6-nitro-3-hydroxybenzaldehyde,2-nitro-5-hydroxybenzaldehyde,5-hydroxy-2-nitro-benzaldehyde,benzaldehyde, 5-hydroxy-2-nitro,unii-va926z93if,6-nitro-3-hydroxy benzaldhyde,pubchem8205,acmc-1an3l,4-08-00-00250 beilstein handbook reference
39211
5-hydroxy-2-nitrobenzaldehyde
C1=CC(=C(C=C1O)C=O)[N+](=O)[O-]
42454-06-8
C7H5NO4
167.12
XLYPHUGUKGMURE-UHFFFAOYSA-N
3-formyl-4-nitrophenol,6-nitro-3-hydroxybenzaldehyde,2-nitro-5-hydroxybenzaldehyde,5-hydroxy-2-nitro-benzaldehyde,benzaldehyde, 5-hydroxy-2-nitro,unii-va926z93if,6-nitro-3-hydroxy benzaldhyde,pubchem8205,acmc-1an3l,4-08-00-00250 beilstein handbook reference
39211
5-hydroxy-2-nitrobenzaldehyde
C1=CC(=C(C=C1O)C=O)[N+](=O)[O-]
Melting Point 168.0°C to 170.0°C
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97.5% min. (GC)
Linear Formula HOC6H3(NO2)CHO
Beilstein 08,63
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde

GHS H Statement
Causes skin irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 255-832-9