Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxy-2-nitrobenzaldehyde, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
42454-06-8
C7H5NO4
167.12
XLYPHUGUKGMURE-UHFFFAOYSA-N
3-formyl-4-nitrophenol,6-nitro-3-hydroxybenzaldehyde,2-nitro-5-hydroxybenzaldehyde,5-hydroxy-2-nitro-benzaldehyde,benzaldehyde, 5-hydroxy-2-nitro,unii-va926z93if,6-nitro-3-hydroxy benzaldhyde,pubchem8205,acmc-1an3l,4-08-00-00250 beilstein handbook reference
39211
5-hydroxy-2-nitrobenzaldehyde
C1=CC(=C(C=C1O)C=O)[N+](=O)[O-]
42454-06-8
C7H5NO4
167.12
XLYPHUGUKGMURE-UHFFFAOYSA-N
3-formyl-4-nitrophenol,6-nitro-3-hydroxybenzaldehyde,2-nitro-5-hydroxybenzaldehyde,5-hydroxy-2-nitro-benzaldehyde,benzaldehyde, 5-hydroxy-2-nitro,unii-va926z93if,6-nitro-3-hydroxy benzaldhyde,pubchem8205,acmc-1an3l,4-08-00-00250 beilstein handbook reference
39211
5-hydroxy-2-nitrobenzaldehyde
C1=CC(=C(C=C1O)C=O)[N+](=O)[O-]
Infrared Spectrum Authentic
Quantity 25g
Melting Point 168.0°C to 170.0°C
Linear Formula HOC6H3(NO2)CHO
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde

GHS H Statement Causes skin irritation. Harmful to aquatic life with long lasting effects.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse. Avoid release to the environment.

Warning