Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Hydroxyindole-3-acetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
1 g
Description
5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.
Solubility
Soluble in ethanol (50 mg/ml).
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
5-Hydroxyindole-3-acetic acid is a major serotonin metabolite.
Solubility
Soluble in ethanol (50 mg/ml).
Notes
Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 161°C to 164°C (decomposition) |
Color | Purple to White |
Quantity | 500 mg |
Beilstein | 168797 |
Sensitivity | Light sensitive |
Solubility Information | Soluble in ethanol (50mg/ml). |
Formula Weight | 191.19 |
Percent Purity | 98% |
Physical Form | Crystalline Powder |
Chemical Name or Material | 5-Hydroxyindole-3-acetic acid |
Safety and Handling
EINECSNumber | 200-195-4 |
RTECSNumber | NL3650000 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction