Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-hmC Mouse anti-Non-species specific, DyLight 650, Clone: 13612, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242219C

Catalog No. NBP242219C


Add to cart

Description

Description

5-hmC Monoclonal antibody specifically detects 5-hmC in Non-species specific samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications

Specifications

5-hmC
13612
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
Mouse
IgG2b
0.1 ml
Primary
56941
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
5'-hmC, 5-hydroxymethylcytosine
The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Other
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit