Learn More
5-hmC Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259182
Catalog No. NBP259182
Add to cart
Description
Description
5-hmC Polyclonal antibody specifically detects 5-hmC in Human, Mouse samples. It is validated for ELISA, Dot Blot, ChIP.
Specifications
Specifications
5-hmC
Unconjugated
Serum with 0.05% Sodium Azide
5'-hmC, 5-hydroxymethylcytosine
Rabbit
IgG
100 ul
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
ELISA, Dot Blot, ChIP
ELISA 1100-12000, Dot Blot 1100-11000, Methylated DNA Immunoprecipitation
Antiserum
HMCES
The exact immunogen is propietary information.
Whole Antiserum
RUO
Primary
56941