Learn More
Aat BioquestSupplier Diversity Partner 5-IAF [5-Iodoacetamidofluorescein] *CAS 63368-54-7* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 222
5-IAF is one of the most popular green fluorescent dyes for thiol modifications of proteins along with fluorescein-5-maleimide. The resulted bioconjugates are quite stable.
Catalog No. 50-195-2814