Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Iodo-1,3-dimethyluracil, 99%

Click to view available options
Quantity:
1g
5g
25g
40738-83-8
C6H7IN2O2
266.038
MFCD00192228
XUUVRIXNFFFPCM-UHFFFAOYSA-N
5-iodo-1,3-dimethyluracil,5-iodo-1,3-dimethylpyrimidine-2,4 1h,3h-dione,5-iodo-1,3-dimethyl-1h-pyrimidine-2,4-dione,acmc-209jgf,1,3-dimethyl-5-iodouracil,1,3-dimethyl 5-iodo uracil,uracil, 1,3-dimethyl-5-iodo,2,4 1h,3h-pyrimidinedione,5-iodo-1,3-dimethyl,5-iodo-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione
622250
5-iodo-1,3-dimethylpyrimidine-2,4-dione
CN1C=C(C(=O)N(C1=O)C)I
40738-83-8
C6H7IN2O2
266.038
MFCD00192228
XUUVRIXNFFFPCM-UHFFFAOYSA-N
5-iodo-1,3-dimethyluracil,5-iodo-1,3-dimethylpyrimidine-2,4 1h,3h-dione,5-iodo-1,3-dimethyl-1h-pyrimidine-2,4-dione,acmc-209jgf,1,3-dimethyl-5-iodouracil,1,3-dimethyl 5-iodo uracil,uracil, 1,3-dimethyl-5-iodo,2,4 1h,3h-pyrimidinedione,5-iodo-1,3-dimethyl,5-iodo-1,3-dimethyl-1,3-dihydropyrimidine-2,4-dione
622250
5-iodo-1,3-dimethylpyrimidine-2,4-dione
CN1C=C(C(=O)N(C1=O)C)I
Melting Point 225°C (decomposition)
Quantity 1g
Formula Weight 266.04
Percent Purity 99%
Chemical Name or Material 5-Iodo-1,3-dimethyluracil

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger