Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Iodo-2,4-dimethoxypyrimidine, 98%

Click to view available options
Quantity:
0.25g
1g
5g
52522-99-3
C6H7IN2O2
266.038
MFCD00090865
KNTMOGOYRYYUIE-UHFFFAOYSA-N
2,4-dimethoxy-5-iodopyrimidine,5-iodo-2,4-dimethoxy-pyrimidine,pyrimidine, 5-iodo-2,4-dimethoxy,pubchem6914,acmc-1apit,5-iodo-2,6-dimethoxypyrimidine,2,4-dimetthoxy-5-iodopyrimidine,5-iodo-2,4-dimethoxypyrimidine,2,4-dimethoxy-5-iodo-pyrimidine,5-iodo-2,4-dimethoxy-pyrimi-dine
263416
5-iodo-2,4-dimethoxypyrimidine
COC1=NC(=NC=C1I)OC
52522-99-3
C6H7IN2O2
266.038
MFCD00090865
KNTMOGOYRYYUIE-UHFFFAOYSA-N
2,4-dimethoxy-5-iodopyrimidine,5-iodo-2,4-dimethoxy-pyrimidine,pyrimidine, 5-iodo-2,4-dimethoxy,pubchem6914,acmc-1apit,5-iodo-2,6-dimethoxypyrimidine,2,4-dimetthoxy-5-iodopyrimidine,5-iodo-2,4-dimethoxypyrimidine,2,4-dimethoxy-5-iodo-pyrimidine,5-iodo-2,4-dimethoxy-pyrimi-dine
263416
5-iodo-2,4-dimethoxypyrimidine
COC1=NC(=NC=C1I)OC
Sensitivity Light sensitive
Quantity 1g
Melting Point 71°C to 75°C
Formula Weight 266.04
Percent Purity 98%
Chemical Name or Material 5-Iodo-2,4-dimethoxypyrimidine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures