Alfa Aesar™ 5-Iodo-2-methoxyaniline, 97%

Manufacturer: Alfa Aesar™ B2555706

Quantity 5g

 View more versions of this product

Catalog No. AAB2555706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 77770-09-3
    Molecular Formula C7H8INO
    Formula Weight 249.05
    MDL Number MFCD03095407
    Synonym 2-Amino-4-iodoanisole; 5-Iodo-o-anisidine
    Chemical Name or Material 5-Iodo-2-methoxyaniline

    Specifications

    Formula Weight 249.05
    Molecular Formula C7H8INO
    MDL Number MFCD03095407
    Melting Point 86° to 87°C
    Synonym 2-Amino-4-iodoanisole; 5-Iodo-o-anisidine
    Quantity 5g
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 77770-09-3
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 5-Iodo-2-methoxyaniline
    Assay 97%

    Description

    Applications
    5-Iodo-2-methoxyaniline is used as a pharmaceutical intermediate.

    Solubility
    Sparingly soluble in water 0.87 g/L @ 25°C.

    Notes
    Light sensitive. Store in dark. Store away from oxidizing agents, light.

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit