Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Iodo-2-methoxybenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
42298-41-9
C8H7IO2
262.046
MFCD04128999
PIRKMHAREMCDPZ-UHFFFAOYSA-N
2-methoxy-5-iodobenzaldehyde,5-iodo-2-methoxy-benzaldehyde,pubchem1445,acmc-20am2z,benzaldehyde,5-iodo-2-methoxy,5-iodo-2-methoxybenzaldehyde
262240
5-iodo-2-methoxybenzaldehyde
COC1=C(C=C(C=C1)I)C=O
42298-41-9
C8H7IO2
262.046
MFCD04128999
PIRKMHAREMCDPZ-UHFFFAOYSA-N
2-methoxy-5-iodobenzaldehyde,5-iodo-2-methoxy-benzaldehyde,pubchem1445,acmc-20am2z,benzaldehyde,5-iodo-2-methoxy,5-iodo-2-methoxybenzaldehyde
262240
5-iodo-2-methoxybenzaldehyde
COC1=C(C=C(C=C1)I)C=O
Quantity 1g
Melting Point 140°C to 146°C
Color White to Yellow
Sensitivity Air & Light Sensitive
Formula Weight 262.04
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-methoxybenzaldehyde