Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodo-2-methylaniline, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
83863-33-6
C7H8IN
233.05
IOEHXNCBPIBDBZ-UHFFFAOYSA-N
2-amino-4-iodotoluene,5-iodo-o-toluidine,5-iodo-2-methyl-phenylamine,benzenamine, 5-iodo-2-methyl,benzenamine,5-iodo-2-methyl,5-iodo-2-methyl-aniline,5-iodo-2-methylbenzenamine,5-iodo-2-methylphenylamine,2-methyl-5-iodoaniline,#
522802
5-iodo-2-methylaniline
CC1=C(C=C(C=C1)I)N
83863-33-6
C7H8IN
233.05
IOEHXNCBPIBDBZ-UHFFFAOYSA-N
2-amino-4-iodotoluene,5-iodo-o-toluidine,5-iodo-2-methyl-phenylamine,benzenamine, 5-iodo-2-methyl,benzenamine,5-iodo-2-methyl,5-iodo-2-methyl-aniline,5-iodo-2-methylbenzenamine,5-iodo-2-methylphenylamine,2-methyl-5-iodoaniline,#
522802
5-iodo-2-methylaniline
CC1=C(C=C(C=C1)I)N
Flash Point >110°C
Melting Point 48°C to 50°C
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-methylaniline

GHS Signal Word: Danger

EINECSNumber : 281-094-2