Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Iodo-A-85380, 5-trimethylstannyl N-BOC derivative, Tocris Bioscience™
Click to view available options
Quantity:
200 μg
Description
Precursor to the highly potent and selective ligand for the α4β2 nicotinic receptor, 5-Iodo-A-85380 (Cat. No.1518). Suitable for radioiodination.
Specifications
Quantity | 200 μg |
Formula Weight | 427.11 |
Percent Purity | >98% |
Chemical Name or Material | 5-Iodo-A-85380, 5-trimethylstannyl N-BOC derivative |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction