Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Iodoindole, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
16066-91-4
C8H6IN
243.04
TVQLYTUWUQMGMP-UHFFFAOYSA-N
5-iodoindole,1h-indole, 5-iodo,5-iodo indole,5-iodanyl-1h-indole,zlchem 1099,acmc-1brxb,maybridge1_006499
2782313
5-iodo-1H-indole
C1=CC2=C(C=CN2)C=C1I
16066-91-4
C8H6IN
243.04
TVQLYTUWUQMGMP-UHFFFAOYSA-N
5-iodoindole,1h-indole, 5-iodo,5-iodo indole,5-iodanyl-1h-indole,zlchem 1099,acmc-1brxb,maybridge1_006499
2782313
5-iodo-1H-indole
C1=CC2=C(C=CN2)C=C1I
Melting Point 95°C to 99°C
Color Brown to White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 243.04
Physical Form Solid or Crystalline Solid
Percent Purity 95%
Chemical Name or Material 5-Iodoindole
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

Warning