Learn More
Medchemexpress LLC 5 Lipoxygenase Antibody (YA1937) | 100 UL

Supplier: Medchemexpress LLC HYP82192100UL
5 Lipoxygenase Antibody (YA1937) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting to 5 Lipoxygenase. It is for research use only. The antibody's host is Rabbit, and it is recombinant, monoclonal. It reacts with Human, Mouse, and Rat species. The immunogen is a synthesized peptide derived from human 5 Lipoxygenase aa120-155. It is supplied in Rabbit IgG in 10mM phosphate buffered saline, pH 7.4, 150mM sodium chloride, 0.05% BSA, 0.02% sodium azide and 50% glycerol.
- Applications: Western Blot (WB), Immunohistochemistry-Paraffin (IHC-P), Immunohistochemistry-Frozen (IHC-F), Immunocytochemistry/Immunofluorescence (ICC/IF), Flow Cytometry (FC)
- Sensitivity: Endogenous
- Conjugation: Non-conjugated
- Isotype: IgG
- Form: Liquid
- Storage: Stored at -20°C for 1 year. Avoid repeated freeze / thaw cycles. Shipping with blue ice.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.