Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid, 96%

Click to view available options
Quantity:
1g
5g
53723-88-9
C4H4N2O2S3
208.268
MFCD00129970
UJBXVTJYSIDCIE-UHFFFAOYSA-N
5-mercapto-1,3,4-thiadiazol-2-ylthio acetic acid,2-5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid,5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid,4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio acetic acid,acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio,5-mercapto-1,3,4-thiadiazole-2-ylthio acetic acid,5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid,5-mercapto-1,3,4-thiadiazole-2-ylthio aceticacid,2-5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid,2-5-sulfanyl-1,3,4-thiadiazol-2-ylthio acetic acid
1714399
2-[(2-sulfanylidene-3H-1,3,4-thiadiazol-5-yl)sulfanyl]acetic acid
C(C(=O)O)SC1=NNC(=S)S1
53723-88-9
C4H4N2O2S3
208.268
MFCD00129970
UJBXVTJYSIDCIE-UHFFFAOYSA-N
5-mercapto-1,3,4-thiadiazol-2-ylthio acetic acid,2-5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid,5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid,4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio acetic acid,acetic acid, 4,5-dihydro-5-thioxo-1,3,4-thiadiazol-2-yl thio,5-mercapto-1,3,4-thiadiazole-2-ylthio acetic acid,5-mercapto-1,3,4-thiadiazol-2-yl thio acetic acid,5-mercapto-1,3,4-thiadiazole-2-ylthio aceticacid,2-5-sulfanyl-1,3,4-thiadiazol-2-yl sulfanyl acetic acid,2-5-sulfanyl-1,3,4-thiadiazol-2-ylthio acetic acid
1714399
2-[(2-sulfanylidene-3H-1,3,4-thiadiazol-5-yl)sulfanyl]acetic acid
C(C(=O)O)SC1=NNC(=S)S1
Melting Point 165°C to 169°C
Quantity 5g
Formula Weight 208.28
Percent Purity 96%
Chemical Name or Material (5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning