Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methoxy-2-methylbenzoselenazole, 97%

Click to view available options
Quantity:
1g
5g
2946-17-0
C9H9NOSe
226.148
MFCD00005558
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methylbenzoselenazole,benzoselenazole, 5-methoxy-2-methyl,5-methoxy-2-methylbenzselenazole,2-methyl-5-methoxybenzoselenazole,5-methoxy-2-methylbenzo d 1,3-selenazole,#,5-methoxy-2-methylbenzo d 1,3 selenazole,5-methoxy-2-methylbenzo d 1,3-selenazoline
76274
5-methoxy-2-methyl-1,3-benzoselenazole
CC1=NC2=C([Se]1)C=CC(=C2)OC
2946-17-0
C9H9NOSe
226.148
MFCD00005558
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methylbenzoselenazole,benzoselenazole, 5-methoxy-2-methyl,5-methoxy-2-methylbenzselenazole,2-methyl-5-methoxybenzoselenazole,5-methoxy-2-methylbenzo d 1,3-selenazole,#,5-methoxy-2-methylbenzo d 1,3 selenazole,5-methoxy-2-methylbenzo d 1,3-selenazoline
76274
5-methoxy-2-methyl-1,3-benzoselenazole
CC1=NC2=C([Se]1)C=CC(=C2)OC
Boiling Point 115°C to 117°C (1mmHg)
Flash Point >110°C (230°F)
Quantity 5g
UN Number UN3440
Formula Weight 226.14
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-methylbenzoselenazole

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: SELENIUM COMPOUND, LIQUID, N.O.S.

TSCA : TSCA