Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-methylbenzoselenazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7026421
Click to view available options
Quantity:
1 g
5 g
2946-17-0
C9H9NOSe
226.148
MFCD00005558
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methylbenzoselenazole, benzoselenazole, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzselenazole, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, #, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline
76274
5-methoxy-2-methyl-1,3-benzoselenazole
CC1=NC2=C([Se]1)C=CC(=C2)OC
2946-17-0
C9H9NOSe
226.148
MFCD00005558
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methylbenzoselenazole, benzoselenazole, 5-methoxy-2-methyl, 5-methoxy-2-methylbenzselenazole, 2-methyl-5-methoxybenzoselenazole, 5-methoxy-2-methylbenzo d 1,3-selenazole, #, 5-methoxy-2-methylbenzo d 1,3 selenazole, 5-methoxy-2-methylbenzo d 1,3-selenazoline
76274
5-methoxy-2-methyl-1,3-benzoselenazole
CC1=NC2=C([Se]1)C=CC(=C2)OC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.49
Boiling Point 115°C to 117°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.639
Quantity 1 g
UN Number UN3440
Formula Weight 226.14
Percent Purity 97%
Chemical Name or Material 5-Methoxy-2-methylbenzoselenazole
Hazard Category H301+H331-H373
Hazard Statement GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement GHS P Statement
P260-P261-P301+P310-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-956-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit