Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-nitrobenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1882-69-5
C8H7NO5
197.15
MFCD00151836
URADKXVAIGMTEG-UHFFFAOYSA-N
5-methoxy-2-nitrobenzoic acid,2-nitro-5-methoxybenzoic acid,6-nitro-m-anisic acid,5-methoxy-2-nitro-benzoic acid,benzoic acid, 5-methoxy-2-nitro,pubchem4229,acmc-1brd6,2-nitro-5-methoxybenzoate,2-nitro-5-methoxybenzoicacid,5-methoxy-2-nitrobenzoicacid
339209
COC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O
1882-69-5
C8H7NO5
197.15
MFCD00151836
URADKXVAIGMTEG-UHFFFAOYSA-N
5-methoxy-2-nitrobenzoic acid,2-nitro-5-methoxybenzoic acid,6-nitro-m-anisic acid,5-methoxy-2-nitro-benzoic acid,benzoic acid, 5-methoxy-2-nitro,pubchem4229,acmc-1brd6,2-nitro-5-methoxybenzoate,2-nitro-5-methoxybenzoicacid,5-methoxy-2-nitrobenzoicacid
339209
COC1=CC(=C(C=C1)[N+](=O)[O-])C(=O)O
Melting Point 125°C to 130°C
Color Beige to Gray or Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 197.15
Physical Form Granules
Percent Purity ≥96.0%
Chemical Name or Material 5-Methoxy-2-nitrobenzoic acid, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.