Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 5-Methoxybenzofuroxan, 97%
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 5g |
Melting Point | 112°C to 114°C |
Beilstein | 643053 |
Formula Weight | 166.14 |
Percent Purity | 97% |
Chemical Name or Material | 5-Methoxybenzofuroxan |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures