Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methoxyflavone, 97%

Click to view available options
Quantity:
0.1g
0.5g
2.5g
42079-78-7
C16H12O3
252.269
MFCD00016942
XRQSPUXANRGDAV-UHFFFAOYSA-N
5-methoxyflavone,5-methoxy-2-phenyl-4h-chromen-4-one,5-methoxy-2-phenyl-4-benzopyrone,chembl16685,4h-1-benzopyran-4-one, 5-methoxy-2-phenyl,5-methoxy-2-phenyl-chromen-4-one,acmc-20am2h,d0v5hf,bidd:er0473
94525
5-methoxy-2-phenylchromen-4-one
COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3
42079-78-7
C16H12O3
252.269
MFCD00016942
XRQSPUXANRGDAV-UHFFFAOYSA-N
5-methoxyflavone,5-methoxy-2-phenyl-4h-chromen-4-one,5-methoxy-2-phenyl-4-benzopyrone,chembl16685,4h-1-benzopyran-4-one, 5-methoxy-2-phenyl,5-methoxy-2-phenyl-chromen-4-one,acmc-20am2h,d0v5hf,bidd:er0473
94525
5-methoxy-2-phenylchromen-4-one
COC1=CC=CC2=C1C(=O)C=C(O2)C3=CC=CC=C3
Melting Point 128°C to 131°C
Quantity 0.1g
Formula Weight 252.27
Percent Purity 97%
Chemical Name or Material 5-Methoxyflavone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning