Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxyindole-2-carboxylic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
4382-54-1
C10H9NO3
191.186
MFCD00005614
YEBJVSLNUMZXRJ-UHFFFAOYSA-N
5-methoxyindole-2-carboxylic acid,1h-indole-2-carboxylic acid, 5-methoxy,5-methoxy-2-indolecarboxylic acid,indole-2-carboxylic acid, 5-methoxy,acide methoxy-5 indole carboxylique-2,acide methoxy-5 indole carboxylique-2 french,5-methoxyindol-2-carboxylic acid,5-methoxy-1h-indole-2-carboxylicacid,pubchem1702,acmc-209jvj
20401
5-methoxy-1H-indole-2-carboxylic acid
COC1=CC2=C(C=C1)NC(=C2)C(=O)O
4382-54-1
C10H9NO3
191.186
MFCD00005614
YEBJVSLNUMZXRJ-UHFFFAOYSA-N
5-methoxyindole-2-carboxylic acid,1h-indole-2-carboxylic acid, 5-methoxy,5-methoxy-2-indolecarboxylic acid,indole-2-carboxylic acid, 5-methoxy,acide methoxy-5 indole carboxylique-2,acide methoxy-5 indole carboxylique-2 french,5-methoxyindol-2-carboxylic acid,5-methoxy-1h-indole-2-carboxylicacid,pubchem1702,acmc-209jvj
20401
5-methoxy-1H-indole-2-carboxylic acid
COC1=CC2=C(C=C1)NC(=C2)C(=O)O
Color Brown to Yellow
Infrared Spectrum Authentic
Melting Point 197°C to 201°C
Quantity 1g
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 22, II, 169
Formula Weight 191.19
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material 5-Methoxyindole-2-carboxylic acid, 99%
Show More Show Less

EINECSNumber : 224-487-6

RTECSNumber : NL6005000