Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxyindole-3-carboxaldehyde 99+%, ACROS Organics™

Click to view available options
Quantity:
100mg
1g
5g
Packaging:
Glass bottle
10601-19-1
C10H9NO2
175.187
MFCD00005623
TUWARWGEOHQXCO-UHFFFAOYSA-N
5-methoxyindole-3-carboxaldehyde,3-formyl-5-methoxyindole,5-methoxyindole-3-carboxyaldehyde,5-methoxyindole-3-carbaldehyde,5-methoxyindole-3-aldehyde,5-methoxy-3-formylindole,1h-indole-3-carboxaldehyde, 5-methoxy,5-methoxy-indol-3-aldehyde,unii-63c50zw2d0,5-methoxy-1h-indole-3-carboxaldehyde
82758
5-methoxy-1H-indole-3-carbaldehyde
COC1=CC2=C(C=C1)NC=C2C=O
10601-19-1
C10H9NO2
175.187
MFCD00005623
TUWARWGEOHQXCO-UHFFFAOYSA-N
5-methoxyindole-3-carboxaldehyde,3-formyl-5-methoxyindole,5-methoxyindole-3-carboxyaldehyde,5-methoxyindole-3-carbaldehyde,5-methoxyindole-3-aldehyde,5-methoxy-3-formylindole,1h-indole-3-carboxaldehyde, 5-methoxy,5-methoxy-indol-3-aldehyde,unii-63c50zw2d0,5-methoxy-1h-indole-3-carboxaldehyde
82758
5-methoxy-1H-indole-3-carbaldehyde
COC1=CC2=C(C=C1)NC=C2C=O
Melting Point 180.0°C to 184.0°C
Color Beige to Yellow
Quantity 100mg
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 21, II, 424
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 175.19
Physical Form Crystalline Powder or Needles
Percent Purity ≥99%
Chemical Name or Material 5-Methoxyindole-3-carboxaldehyde, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 234-220-5