Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methoxyindole-3-carboxylic acid, 97%

Click to view available options
Quantity:
250mg
1g
5g
10242-01-0
C10H9NO3
191.186
MFCD03265451
RVVSEZGJCOAUED-UHFFFAOYSA-N
5-methoxyindole-3-carboxylic acid,5-methoxy-3-indolecarboxylic acid,5-methoxy-3-carboxyindole,1h-indole-3-carboxylic acid, 5-methoxy,pubchem7984,zlchem 1276,1h-indole-3-carboxylicacid, 5-methoxy,5-methoxy-3-indolecarboxylic acid,5-methoxyindole-3-carboxylic acid;,5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
259191
5-methoxy-1H-indole-3-carboxylic acid
COC1=CC2=C(C=C1)NC=C2C(=O)O
10242-01-0
C10H9NO3
191.186
MFCD03265451
RVVSEZGJCOAUED-UHFFFAOYSA-N
5-methoxyindole-3-carboxylic acid,5-methoxy-3-indolecarboxylic acid,5-methoxy-3-carboxyindole,1h-indole-3-carboxylic acid, 5-methoxy,pubchem7984,zlchem 1276,1h-indole-3-carboxylicacid, 5-methoxy,5-methoxy-3-indolecarboxylic acid,5-methoxyindole-3-carboxylic acid;,5-methoxyindole-3-carboxylic acid 5-methoxy-1h-indazole-3-carboxylic acid
259191
5-methoxy-1H-indole-3-carboxylic acid
COC1=CC2=C(C=C1)NC=C2C(=O)O
Quantity 5g
Color Red
Beilstein 475711
Formula Weight 191.19
Physical Form Crystalline
Percent Purity 97%
Chemical Name or Material 5-Methoxyindole-3-carboxylic acid

storageNote1 : 2° to 8°C