Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxyindole 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1006-94-6
C9H9NO
147.18
MFCD00005674
DWAQDRSOVMLGRQ-UHFFFAOYSA-N
5-methoxyindole,1h-indole, 5-methoxy,femedol,5-methoxy indole,indol-5-yl methyl ether,indole, 5-methoxy,methoxy-5 indole,unii-dqm3as43pq,methoxy-5 indole french,methoxyindole 5-
13872
5-methoxy-1H-indole
COC1=CC2=C(C=C1)NC=C2
1006-94-6
C9H9NO
147.18
MFCD00005674
DWAQDRSOVMLGRQ-UHFFFAOYSA-N
5-methoxyindole,1h-indole, 5-methoxy,femedol,5-methoxy indole,indol-5-yl methyl ether,indole, 5-methoxy,methoxy-5 indole,unii-dqm3as43pq,methoxy-5 indole french,methoxyindole 5-
13872
5-methoxy-1H-indole
COC1=CC2=C(C=C1)NC=C2
Quantity 5g
Infrared Spectrum Authentic
Color Brown to White
Melting Point 52.0°C to 56.0°C
Boiling Point 176.0°C to 178.0°C (17.0mmHg)
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 21, II, 44
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone
Formula Weight 147.18
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 5-Methoxyindole, 99%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 213-745-3

RTECSNumber : NL9500000