Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxyisatin, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
100g
39755-95-8
C9H7NO3
177.16
DMHGXMPXHPOXBF-UHFFFAOYSA-N
5-methoxyisatin,5-methoxyindoline-2,3-dione,5-methoxy-2,3-indolinedione,1h-indole-2,3-dione, 5-methoxy,5-methoxy-indole-2,3-dione,5-methoxy isatin,5-methoxyindole-2,3-dione,5-methoxy-2,3-dihydro-1h-indole-2,3-dione,indole-2,3-dione, 5-methoxy,1h-indole-2,3-dione, 5-methoxy-9ci
38333
5-methoxy-1H-indole-2,3-dione
COC1=CC2=C(C=C1)NC(=O)C2=O
39755-95-8
C9H7NO3
177.16
DMHGXMPXHPOXBF-UHFFFAOYSA-N
5-methoxyisatin,5-methoxyindoline-2,3-dione,5-methoxy-2,3-indolinedione,1h-indole-2,3-dione, 5-methoxy,5-methoxy-indole-2,3-dione,5-methoxy isatin,5-methoxyindole-2,3-dione,5-methoxy-2,3-dihydro-1h-indole-2,3-dione,indole-2,3-dione, 5-methoxy,1h-indole-2,3-dione, 5-methoxy-9ci
38333
5-methoxy-1H-indole-2,3-dione
COC1=CC2=C(C=C1)NC(=O)C2=O
Melting Point 201°C to 203°C
Packaging Glass bottle
Quantity 100g
Assay Percent Range 96% min. (GC)
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-Methoxyisatin

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning