Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methoxyisatin, 98%

Click to view available options
Quantity:
5g
25g
100g
39755-95-8
C9H7NO3
177.159
MFCD00169023
DMHGXMPXHPOXBF-UHFFFAOYSA-N
5-methoxyisatin,5-methoxyindoline-2,3-dione,5-methoxy-2,3-indolinedione,1h-indole-2,3-dione, 5-methoxy,5-methoxy-indole-2,3-dione,5-methoxy isatin,5-methoxyindole-2,3-dione,5-methoxy-2,3-dihydro-1h-indole-2,3-dione,indole-2,3-dione, 5-methoxy,1h-indole-2,3-dione, 5-methoxy-9ci
38333
5-methoxy-1H-indole-2,3-dione
COC1=CC2=C(C=C1)NC(=O)C2=O
39755-95-8
C9H7NO3
177.159
MFCD00169023
DMHGXMPXHPOXBF-UHFFFAOYSA-N
5-methoxyisatin,5-methoxyindoline-2,3-dione,5-methoxy-2,3-indolinedione,1h-indole-2,3-dione, 5-methoxy,5-methoxy-indole-2,3-dione,5-methoxy isatin,5-methoxyindole-2,3-dione,5-methoxy-2,3-dihydro-1h-indole-2,3-dione,indole-2,3-dione, 5-methoxy,1h-indole-2,3-dione, 5-methoxy-9ci
38333
5-methoxy-1H-indole-2,3-dione
COC1=CC2=C(C=C1)NC(=O)C2=O
Applications
5-Methoxyisatin is used for Pharmaceutical intermediates, a raw material used in chemical synthesis.

Solubility
Soluble in cold water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Melting Point 199°C to 202°C
Quantity 100g
Formula Weight 177.16
Percent Purity 98%
Chemical Name or Material 5-Methoxyisatin

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NL7938000