Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methoxypyrazine-2-carboxylic acid, 98%

Click to view available options
Quantity:
250mg
1g
40155-42-8
C6H6N2O3
154.125
MFCD10697773
YVGVOPNUEFTVQO-UHFFFAOYSA-N
2-carboxy-5-methoxypyrazine,2-carboxy-5-methoxy-1,4-diazine,2-pyrazinecarboxylicacid, 5-methoxy,5-methoxy-2-pyrazinecarboxylic acid,2-pyrazinecarboxylic acid, 5-methoxy,5-methoxy-pyrazine-2-carboxylic acid,5-methyloxy-2-pyrazinecarboxylic acid,5-methoxy-pyrazinecarboxylic acid,5-methoxypyrazine-2-carboxylic acpubchem_ext_datasource_regid
45789811
5-methoxypyrazine-2-carboxylic acid
COC1=NC=C(N=C1)C(=O)O
40155-42-8
C6H6N2O3
154.125
MFCD10697773
YVGVOPNUEFTVQO-UHFFFAOYSA-N
2-carboxy-5-methoxypyrazine,2-carboxy-5-methoxy-1,4-diazine,2-pyrazinecarboxylicacid, 5-methoxy,5-methoxy-2-pyrazinecarboxylic acid,2-pyrazinecarboxylic acid, 5-methoxy,5-methoxy-pyrazine-2-carboxylic acid,5-methyloxy-2-pyrazinecarboxylic acid,5-methoxy-pyrazinecarboxylic acid,5-methoxypyrazine-2-carboxylic acpubchem_ext_datasource_regid
45789811
5-methoxypyrazine-2-carboxylic acid
COC1=NC=C(N=C1)C(=O)O
Quantity 250mg
Beilstein 908499
Formula Weight 154.13
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Methoxypyrazine-2-carboxylic acid

storageNote1 : 2° to 8°C