Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxypyridine-3-boronic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
850991-69-4
C6H8BNO3
152.95
MFCD03425977
ISDFOFZTZUILPE-UHFFFAOYSA-N
5-methoxypyridine-3-boronic acid,5-methoxypyridin-3-yl boronic acid,3-methoxy-5-pyridineboronic acid,5-methoxy-3-pyridyl boronic acid,3-borono-5-methoxypyridine,5-methoxy-3-pyridinyl boronic acid,3-methoxypyridine-5-boronic acid,5-methoxy-3-pyridinylboronic acid,5-methoxy-3-pyridineboronic acid,boronic acid,b-5-methoxy-3-pyridinyl
2763087
(5-methoxypyridin-3-yl)boronic acid
B(C1=CC(=CN=C1)OC)(O)O
850991-69-4
C6H8BNO3
152.95
MFCD03425977
ISDFOFZTZUILPE-UHFFFAOYSA-N
5-methoxypyridine-3-boronic acid,5-methoxypyridin-3-yl boronic acid,3-methoxy-5-pyridineboronic acid,5-methoxy-3-pyridyl boronic acid,3-borono-5-methoxypyridine,5-methoxy-3-pyridinyl boronic acid,3-methoxypyridine-5-boronic acid,5-methoxy-3-pyridinylboronic acid,5-methoxy-3-pyridineboronic acid,boronic acid,b-5-methoxy-3-pyridinyl
2763087
(5-methoxypyridin-3-yl)boronic acid
B(C1=CC(=CN=C1)OC)(O)O
Quantity 1g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 152.95
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 5-Methoxypyridine-3-boronic acid
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning