Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxytryptamine Hydrochloride 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
66-83-1
C11H14N2O·HCl
226.71
MFCD00012684
TXVAYRSEKRMEIF-UHFFFAOYSA-N
5-methoxytryptamine hydrochloride,mexamine hydrochloride,o-methylserotonin hydrochloride,2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride,3-2-aminoethyl-5-methoxyindole hydrochloride,unii-4265ib6qek,5-methoxy-1h-indole-3-ethylamine hydrochloride,1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride,5-methoxy tryptamine hcl,5 methoxytryptamine hydrochloride
6198
2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
COC1=CC2=C(C=C1)NC=C2CCN.Cl
66-83-1
C11H14N2O·HCl
226.71
MFCD00012684
TXVAYRSEKRMEIF-UHFFFAOYSA-N
5-methoxytryptamine hydrochloride,mexamine hydrochloride,o-methylserotonin hydrochloride,2-5-methoxy-1h-indol-3-yl ethanamine hydrochloride,3-2-aminoethyl-5-methoxyindole hydrochloride,unii-4265ib6qek,5-methoxy-1h-indole-3-ethylamine hydrochloride,1h-indole-3-ethanamine, 5-methoxy-, monohydrochloride,5-methoxy tryptamine hcl,5 methoxytryptamine hydrochloride
6198
2-(5-methoxy-1H-indol-3-yl)ethanamine;hydrochloride
COC1=CC2=C(C=C1)NC=C2CCN.Cl
Color Beige to White
Infrared Spectrum Authentic
Melting Point 246.0°C to 248.0°C
Quantity 250mg
Water 0.5% max.
Loss on Drying 1% max.
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Merck Index 14, 6002
Formula Weight 226.71
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Methoxytryptamine hydrochloride, 97%
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-637-6

RTECSNumber : NL4110000