Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-1,3-oxazole-4-carboxylic acid, 95%, Acros Organics™

Click to view available options
Quantity:
1g
5g
103879-58-9
C5H5NO3
127.1
QIACATCUODRSLS-UHFFFAOYSA-N
5-methyloxazole-4-carboxylic acid,4-oxazolecarboxylic acid, 5-methyl,5-methyloxazole-4-carboxylic ac,4-oxazolecarboxylicacid, 5-methyl,5-methyl-oxazole-4-carboxylic acid,acmc-209zwx,5-methyloxazole-4-carboxylicacid,5-methyl-4-oxazolecarboxylic acid,4-carboxy-5-methyl-1,3-oxazole,5-methyl oxazole-4-carboxylic acid
11355484
5-methyl-1,3-oxazole-4-carboxylic acid
CC1=C(N=CO1)C(=O)O
103879-58-9
C5H5NO3
127.1
QIACATCUODRSLS-UHFFFAOYSA-N
5-methyloxazole-4-carboxylic acid,4-oxazolecarboxylic acid, 5-methyl,5-methyloxazole-4-carboxylic ac,4-oxazolecarboxylicacid, 5-methyl,5-methyl-oxazole-4-carboxylic acid,acmc-209zwx,5-methyloxazole-4-carboxylicacid,5-methyl-4-oxazolecarboxylic acid,4-carboxy-5-methyl-1,3-oxazole,5-methyl oxazole-4-carboxylic acid
11355484
5-methyl-1,3-oxazole-4-carboxylic acid
CC1=C(N=CO1)C(=O)O
Packaging Glass Bottle
Melting Point 149.0°C to 153.0°C
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 94% min. (HPLC)
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 5-Methyl-1,3-oxazole-4-carboxylic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning