Alfa Aesar™ 5-Methyl-1-hexyn-3-ol, 97%

Manufacturer: Alfa Aesar™ H5349006

Quantity 5g

 View more versions of this product

Catalog No. AAH5349006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 61996-79-0
    Molecular Formula C7H12O
    Formula Weight 112.17
    MDL Number MFCD00041606
    UN Number UN1987
    Beilstein 1741083
    Chemical Name or Material 5-Methyl-1-hexyn-3-ol

    Specifications

    Formula Weight 112.17
    Molecular Formula C7H12O
    MDL Number MFCD00041606
    CAS 61996-79-0
    UN Number UN1987
    Beilstein 1741083
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H226
    Toxic if swallowed.
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
    Flash Point 53°C (127°F)
    Quantity 5g
    Chemical Name or Material 5-Methyl-1-hexyn-3-ol
    Assay 97%

    GHS H Statement
    H301-H226
    Toxic if swallowed.
    Flammable liquid and vapor.

    2° to 8°C

    GHS P Statement
    P210-P280-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit