Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
894779-67-0
C11H13N3
187.24
GRLXUIRJMGNTQO-UHFFFAOYSA-N
5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine,1h-pyrazole-4-methanamine, 5-methyl-1-phenyl,5-methyl-1-phenylpyrazol-4-yl methanamine,5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine
28472963
(5-methyl-1-phenylpyrazol-4-yl)methanamine
CC1=C(C=NN1C2=CC=CC=C2)CN
894779-67-0
C11H13N3
187.24
GRLXUIRJMGNTQO-UHFFFAOYSA-N
5-methyl-1-phenyl-1h-pyrazol-4-yl methanamine,1h-pyrazole-4-methanamine, 5-methyl-1-phenyl,5-methyl-1-phenylpyrazol-4-yl methanamine,5-methyl-1-phenyl-1h-pyrazol-4-yl methyl amine
28472963
(5-methyl-1-phenylpyrazol-4-yl)methanamine
CC1=C(C=NN1C2=CC=CC=C2)CN
Quantity 5g
Color Colorless to Yellow or White to Yellow
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (HPLC)
Formula Weight 187.24
Physical Form Liquid or Powder
Percent Purity 95%
Chemical Name or Material 5-Methyl-1-phenyl-(1H-pyrazol-4-ylmethyl)amine

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger