Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-1H-benzotriazole, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
136-85-6
C7H7N3
133.15
MFCD00005702
LRUDIIUSNGCQKF-UHFFFAOYSA-N
5-methyl-1h-benzotriazole,5-methylbenzotriazole,tolutriazole,5-tolyltriazole,1h-benzotriazole, 5-methyl,6-methylbenzotriazole,5-methyl-1h-benzo d 1,2,3 triazole,5-methyl-1,2,3-benzotriazole,6-methyl-1,2,3-benzotriazole,t56 bmnnj g1
8705
CHEBI:83455
5-methyl-2H-benzotriazole
CC1=CC2=NNN=C2C=C1
136-85-6
C7H7N3
133.15
MFCD00005702
LRUDIIUSNGCQKF-UHFFFAOYSA-N
5-methyl-1h-benzotriazole,5-methylbenzotriazole,tolutriazole,5-tolyltriazole,1h-benzotriazole, 5-methyl,6-methylbenzotriazole,5-methyl-1h-benzo d 1,2,3 triazole,5-methyl-1,2,3-benzotriazole,6-methyl-1,2,3-benzotriazole,t56 bmnnj g1
8705
CHEBI:83455
5-methyl-2H-benzotriazole
CC1=CC2=NNN=C2C=C1
Boiling Point 210.0°C to 212.0°C (12.0mmHg)
Infrared Spectrum Authentic
Melting Point 80.0°C to 84.0°C
Color Cream to Beige
Quantity 10g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Beilstein 26, 58
Solubility Information Solubility in water: 6.0g/L (25 c). Other solubilities: soluble in acids and alkalies,soluble in methanol and ethanol
Formula Weight 133.15
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material 5-Methyl-1H-benzotriazole, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 205-265-8

RTECSNumber : DM1400000

TSCA : TSCA