Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-(2,2,2-trimethylacetamido)pyridine, 98%

Click to view available options
Quantity:
250mg
1g
5g
86847-78-1
C11H16N2O
192.262
MFCD03374646
XUCQZJBNEVIURX-UHFFFAOYSA-N
n-5-methylpyridin-2-yl pivalamide,2,2-dimethyl-n-5-methylpyridin-2-yl propanamide,2,2-dimethyl-n-5-methyl-pyridin-2-yl-propionamide,6-pivaloylamino 3-picoline,n-5-methyl-2-pyridinyl pivalamide,2,2-dimethyl-n-5-methyl-2-pyridinyl propanamide,propanamide, 2,2-dimethyl-n-5-methyl-2-pyridinyl
4182924
2,2-dimethyl-N-(5-methylpyridin-2-yl)propanamide
CC1=CN=C(C=C1)NC(=O)C(C)(C)C
86847-78-1
C11H16N2O
192.262
MFCD03374646
XUCQZJBNEVIURX-UHFFFAOYSA-N
n-5-methylpyridin-2-yl pivalamide,2,2-dimethyl-n-5-methylpyridin-2-yl propanamide,2,2-dimethyl-n-5-methyl-pyridin-2-yl-propionamide,6-pivaloylamino 3-picoline,n-5-methyl-2-pyridinyl pivalamide,2,2-dimethyl-n-5-methyl-2-pyridinyl propanamide,propanamide, 2,2-dimethyl-n-5-methyl-2-pyridinyl
4182924
2,2-dimethyl-N-(5-methylpyridin-2-yl)propanamide
CC1=CN=C(C=C1)NC(=O)C(C)(C)C
Quantity 1g
Beilstein 4179181
Formula Weight 192.26
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 5-Methyl-2-(2,2,2-trimethylacetamido)pyridine