Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-(2-thienyl)benzimidazole, 95%

Click to view available options
Quantity:
250mg
1g
5g
3878-24-8
C12H10N2S
214.286
MFCD06312144
QIYVOSKFSQXPCG-UHFFFAOYSA-N
6-methyl-2-2-thienyl benzimidazole,5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole,5-methyl-2-thiophen-2-yl-1h-benzoimidazole,5-methyl-2-2-thienyl benzimidazole,2-5-methylbenzimidazol-2-yl thiophene,5-methyl-2-thien-2-yl-1h-benzimidazole,2-2-thienyl-5-methyl-1h-benzimidazole,5-methyl-2-thiophen-2-yl-1h-benzimidazole,5-methyl-2-thiophen-2-yl-1h-benzo d imidazole,6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
924806
6-methyl-2-thiophen-2-yl-1H-benzimidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3
3878-24-8
C12H10N2S
214.286
MFCD06312144
QIYVOSKFSQXPCG-UHFFFAOYSA-N
6-methyl-2-2-thienyl benzimidazole,5-methyl-2-thiophen-2-yl-3h-1,3-benzodiazole,5-methyl-2-thiophen-2-yl-1h-benzoimidazole,5-methyl-2-2-thienyl benzimidazole,2-5-methylbenzimidazol-2-yl thiophene,5-methyl-2-thien-2-yl-1h-benzimidazole,2-2-thienyl-5-methyl-1h-benzimidazole,5-methyl-2-thiophen-2-yl-1h-benzimidazole,5-methyl-2-thiophen-2-yl-1h-benzo d imidazole,6-methyl-2-thiophen-2-yl-1h-benzo d imidazole
924806
6-methyl-2-thiophen-2-yl-1H-benzimidazole
CC1=CC2=C(C=C1)N=C(N2)C3=CC=CS3
Quantity 5g
Formula Weight 214.29
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Methyl-2-(2-thienyl)benzimidazole