Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-(4-methylphenyl)benzimidazole, 95%

Click to view available options
Quantity:
250mg
1g
5g
7118-66-3
C15H14N2
222.291
MFCD00436743
OPBSAVHQPYKEQP-UHFFFAOYSA-N
5-methyl-2-p-tolyl-1h-benzoimidazole,5-methyl-2-4-methylphenyl-1h-benzimidazole,6-methyl-2-4-methylphenyl-1h-benzimidazole,5-methyl-2-4-methylphenyl benzimidazole,5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole,5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole,2-4-methylphenyl-5-methyl-1h-benzimidazole,1h-benzimidazole, 5-methyl-2-4-methylphenyl
893072
6-methyl-2-(4-methylphenyl)-1H-benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C
7118-66-3
C15H14N2
222.291
MFCD00436743
OPBSAVHQPYKEQP-UHFFFAOYSA-N
5-methyl-2-p-tolyl-1h-benzoimidazole,5-methyl-2-4-methylphenyl-1h-benzimidazole,6-methyl-2-4-methylphenyl-1h-benzimidazole,5-methyl-2-4-methylphenyl benzimidazole,5-methyl-2-4-methylphenyl-1h-1,3-benzodiazole,5-methyl-2-4-methylphenyl-3h-1,3-benzodiazole,2-4-methylphenyl-5-methyl-1h-benzimidazole,1h-benzimidazole, 5-methyl-2-4-methylphenyl
893072
6-methyl-2-(4-methylphenyl)-1H-benzimidazole
CC1=CC=C(C=C1)C2=NC3=C(N2)C=C(C=C3)C
Quantity 5g
Formula Weight 222.29
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 5-Methyl-2-(4-methylphenyl)benzimidazole