Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Methyl-2-furoyl chloride, 97%

Manufacturer:  Alfa Aesar™ H2769703

Quantity 1g

 View more versions of this product

Catalog No. AAH2769703


  Check Availability
Add to cart

Specifications

Description

Applications
As pharmaceutical intermediates and reagent. In chemical research. The intermediate 2-halo- acetylfurans (chloro or bromo) were obtained by a hazardous procedure involving the treatment of 2-furoylchloride (highly lachrymatory) with diazomethane followed by the decomposition of the intermediate with dry HCl. Three monoprotic tridentate ligands were synthesized by condensation of 2-furoylchloride with anthranilic acid and β-alanine, and 2-acetylpyridine with orthanilic acid.

Solubility
Reacts with water.

Notes
Highly lachrymatory. Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases, water, moisture.
Description & Specifications

Chemical Identifiers

14003-11-3
144.56
UN3261
5-Methyl-2-furoyl chloride
C6H5ClO2
MFCD03421416
5-Methylfuran-2-carbonyl chloride

Specifications

14003-11-3
144.56
UN3261
27° to 30°C
91°C (196°F)
97%
C6H5ClO2
MFCD03421416
5-Methylfuran-2-carbonyl chloride
1g
5-Methyl-2-furoyl chloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit